Product Details

SNP ID
rs2277927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32597460 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAAAAAGGGATGTTTCTAGAAA[C/T]ACTGCTACAGGTGCACGTGTAGTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032139.2 3994 UTR 3 NP_115515.2
XM_006723412.2 3994 UTR 3 XP_006723475.1
XM_017027349.1 3994 UTR 3 XP_016882838.1

View Full Product Details