Product Details
- SNP ID
-
rs2981977
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:167336662 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCGAGACAAGACAGTGGTTTTGCTC[A/G]TTGGCTCAGCTGTCCTGTTCCAGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTLL2
PubMed Links
Gene Details
- Gene
- TTLL2
- Gene Name
- tubulin tyrosine ligase like 2
View Full Product Details