Product Details
- SNP ID
-
rs2291379
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:146255419 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGCATCCATGACCTTTATATTTT[A/C]ATTTATTCATGAAAGCAATATGATT
- Phenotype
-
MIM: 611459
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC10A7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79938523] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC10A7
- Gene Name
- solute carrier family 10 member 7
View Full Product Details