Product Details

SNP ID
rs2291382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131469684 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTGTTCCTAAAAAGAACCACT[G/T]GTAACACTTACTTTCAGTCCATATT
Phenotype
MIM: 607118
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL3 PubMed Links

Gene Details

Gene
MRPL3
Gene Name
mitochondrial ribosomal protein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007208.3 Intron NP_009139.1
Gene
SNORA58
Gene Name
small nucleolar RNA, H/ACA box 58
There are no transcripts associated with this gene.

View Full Product Details