Product Details

SNP ID
rs2291381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131469730 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTCTGTCCTGTATATGTTTCCC[A/G]TTTTTCCAGGCATTTTAGTTCCAGG
Phenotype
MIM: 607118
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL3 PubMed Links

Gene Details

Gene
MRPL3
Gene Name
mitochondrial ribosomal protein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007208.3 977 Missense Mutation ACG,ATG T261M NP_009139.1
Gene
SNORA58
Gene Name
small nucleolar RNA, H/ACA box 58
There are no transcripts associated with this gene.

View Full Product Details