Product Details

SNP ID
rs2291457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130270623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACTGGGGACCCGAGTGTTACC[C/G]TTTCCTTCACAATTACAGACTGAAA
Phenotype
MIM: 600688
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPA2 PubMed Links

Gene Details

Gene
CPA2
Gene Name
carboxypeptidase A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001869.2 Intron NP_001860.2

View Full Product Details