Product Details
- SNP ID
-
rs2291725
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48961770 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCTCCTCCTTCCTATTAGCTTGAC[C/T]GGCCAGCTCCAGCGCCCGAGCCTCC
- Phenotype
-
MIM: 137240
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GIP
PubMed Links
Gene Details
- Gene
- GIP
- Gene Name
- gastric inhibitory polypeptide
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004123.2 |
405 |
Missense Mutation |
AGT,GGT |
S103G |
NP_004114.1 |
View Full Product Details