Product Details

SNP ID
rs2291746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98224533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCCTGGCACAGCAGGGATGAAC[A/G]TGGCGTGGGGGAGCCATGGTGCTCC
Phenotype
MIM: 614781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TECPR1 PubMed Links

Gene Details

Gene
TECPR1
Gene Name
tectonin beta-propeller repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015395.2 Intron NP_056210.1
XM_005250253.3 Intron XP_005250310.1
XM_017011937.1 Intron XP_016867426.1
XM_017011938.1 Intron XP_016867427.1

View Full Product Details