Product Details

SNP ID
rs2291841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5689666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGCCAGTACAGCAGATGCTAGA[A/C]CATAATGTAGCATTACTTTCCCCAG
Phenotype
MIM: 606559 MIM: 608487
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRIM22 PubMed Links

Gene Details

Gene
TRIM22
Gene Name
tripartite motif containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199573.1 80 UTR 5 NP_001186502.1
NM_006074.4 80 UTR 5 NP_006065.2
Gene
TRIM5
Gene Name
tripartite motif containing 5
There are no transcripts associated with this gene.

View Full Product Details