Product Details
- SNP ID
-
rs2292201
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:49298568 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCATGAGCCCACCTCAGGGTTGAA[G/T]GGAGGCCAGCCCTTCCCAGAGGTGG
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C10orf71
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75022241] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C10orf71
- Gene Name
- chromosome 10 open reading frame 71
- Gene
- C10orf71-AS1
- Gene Name
- C10orf71 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details