Product Details
- SNP ID
-
rs2292276
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
75
- Location
-
Chr.1:163322572 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GCATGCATAACGCCATCATTGTTCC[C/T]TTTAAAAGTTCAAGGAACGTTTTAC
- Phenotype
-
MIM: 611772
MIM: 603276
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NUF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12755500] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NUF2
- Gene Name
- NUF2, NDC80 kinetochore complex component
- Gene
- RGS5
- Gene Name
- regulator of G-protein signaling 5
View Full Product Details