Product Details

SNP ID
rs2279516
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:53560674 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGCCTGCAGCAAGATGGCGGCGGT[G/C]TCAATGTCAGTGGTACTGAGGCAGA
Phenotype
MIM: 602694
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFS4 PubMed Links

Gene Details

Gene
NDUFS4
Gene Name
NADH:ubiquinone oxidoreductase subunit S4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318051.1 42 Silent Mutation GTC,GTG V4V NP_001304980.1
NM_002495.3 42 Silent Mutation GTC,GTG V4V NP_002486.1
XM_017009491.1 42 Silent Mutation GTC,GTG V4V XP_016864980.1

View Full Product Details