Product Details
- SNP ID
-
rs2292450
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1525383 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCCTCAAGGTGGTGCCGTGTGGC[A/G]GGGCTGGGGCCGGGTGGCTTCGCCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLK5
PubMed Links
Gene Details
- Gene
- PLK5
- Gene Name
- polo like kinase 5
View Full Product Details