Product Details

SNP ID
rs2279695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116716457 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGACCTTGCTCAATCCTGAGGTT[C/T]GTGAGGAATATAGGAGAGTTGTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf49 PubMed Links

Gene Details

Gene
C12orf49
Gene Name
chromosome 12 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024738.2 2141 UTR 3 NP_079014.1
XM_005253937.3 2141 UTR 3 XP_005253994.1

View Full Product Details