Product Details
- SNP ID
-
rs2279819
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126007205 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGTCAGCCGAACCATCACTTCTGC[C/G]AGGTACAGCAGGATTGTCACCTGTC
- Phenotype
-
MIM: 610803
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC41A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79696429] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC41A3
- Gene Name
- solute carrier family 41 member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008485.1 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
NP_001008485.1 |
NM_001008486.1 |
1424 |
Silent Mutation |
CTC,CTG |
L389L |
NP_001008486.1 |
NM_001008487.1 |
1424 |
Silent Mutation |
CTC,CTG |
L399L |
NP_001008487.1 |
NM_001164475.1 |
1424 |
Silent Mutation |
CTC,CTG |
L308L |
NP_001157947.1 |
NM_017836.3 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
NP_060306.3 |
XM_005247559.1 |
1424 |
Silent Mutation |
CTC,CTG |
L440L |
XP_005247616.1 |
XM_005247562.1 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
XP_005247619.1 |
XM_005247563.2 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
XP_005247620.1 |
XM_005247564.1 |
1424 |
Silent Mutation |
CTC,CTG |
L440L |
XP_005247621.1 |
XM_005247565.2 |
1424 |
Silent Mutation |
CTC,CTG |
L399L |
XP_005247622.1 |
XM_006713681.2 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
XP_006713744.1 |
XM_011512945.1 |
1424 |
Silent Mutation |
CTC,CTG |
L414L |
XP_011511247.1 |
XM_017006705.1 |
1424 |
Silent Mutation |
CTC,CTG |
L440L |
XP_016862194.1 |
XM_017006706.1 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
XP_016862195.1 |
XM_017006707.1 |
1424 |
Silent Mutation |
CTC,CTG |
L425L |
XP_016862196.1 |
XM_017006708.1 |
1424 |
Silent Mutation |
CTC,CTG |
L389L |
XP_016862197.1 |
XM_017006709.1 |
1424 |
Silent Mutation |
CTC,CTG |
L389L |
XP_016862198.1 |
View Full Product Details