Product Details
- SNP ID
-
rs2279841
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:15601668 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGAGGTGAGAATGTAAACACGCGC[A/G]TTCTCCAATCAGAACTGCGCTCTCT
- Phenotype
-
MIM: 609019
MIM: 604300
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115586700] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTD
- Gene Name
- biotinidase
- Gene
- HACL1
- Gene Name
- 2-hydroxyacyl-CoA lyase 1
View Full Product Details