Product Details

SNP ID
rs2292685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103748892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACACCACTAGTGCTGTGGTGCC[C/T]CATACCCTGACCTGAAGCCCTAGTT
Phenotype
MIM: 611076 MIM: 608561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NT5DC3 PubMed Links

Gene Details

Gene
NT5DC3
Gene Name
5'-nucleotidase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031701.2 Intron NP_001026871.1
XM_011538476.2 Intron XP_011536778.1
XM_017019454.1 Intron XP_016874943.1
Gene
STAB2
Gene Name
stabilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017564.9 Intron NP_060034.9
XM_011538537.2 Intron XP_011536839.1
XM_011538538.2 Intron XP_011536840.1
XM_011538539.2 Intron XP_011536841.1
XM_011538541.2 Intron XP_011536843.1
XM_011538542.2 Intron XP_011536844.1
XM_017019585.1 Intron XP_016875074.1

View Full Product Details