Product Details

SNP ID
rs2292720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49990388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTGGAAAAATATTCTATAAAAAA[C/T]GGTTGAATAAACAACTATAATATTA
Phenotype
MIM: 604980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RACGAP1 PubMed Links

Gene Details

Gene
RACGAP1
Gene Name
Rac GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126103.2 Intron NP_001119575.1
NM_001126104.2 Intron NP_001119576.1
NM_001319999.1 Intron NP_001306928.1
NM_001320000.1 Intron NP_001306929.1
NM_001320001.1 Intron NP_001306930.1
NM_001320002.1 Intron NP_001306931.1
NM_001320003.1 Intron NP_001306932.1
NM_001320004.1 Intron NP_001306933.1
NM_001320005.1 Intron NP_001306934.1
NM_001320006.1 Intron NP_001306935.1
NM_001320007.1 Intron NP_001306936.1
NM_013277.4 Intron NP_037409.2
XM_006719359.1 Intron XP_006719422.1
XM_011538238.1 Intron XP_011536540.1
XM_011538243.1 Intron XP_011536545.1
XM_017019220.1 Intron XP_016874709.1
XM_017019221.1 Intron XP_016874710.1
XM_017019222.1 Intron XP_016874711.1
XM_017019223.1 Intron XP_016874712.1
XM_017019224.1 Intron XP_016874713.1
XM_017019225.1 Intron XP_016874714.1
XM_017019226.1 Intron XP_016874715.1
XM_017019227.1 Intron XP_016874716.1

View Full Product Details