Product Details

SNP ID
rs2279996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113737728 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTAGAAGAATACAGCTATTTAC[A/G]TGGAAGAAAGATTACTGTGACTCGT
Phenotype
MIM: 603954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZW10 PubMed Links

Gene Details

Gene
ZW10
Gene Name
zw10 kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004724.3 Intron NP_004715.1
XM_017018558.1 Intron XP_016874047.1
XM_017018559.1 Intron XP_016874048.1

View Full Product Details