Product Details

SNP ID
rs2292835
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:96231492 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTTTTCATCTAAGTAGGAAGCAGA[A/G]TGCAGAGCTGGAACAGCAAAGACCT
Phenotype
MIM: 616930 MIM: 191330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTERF3 PubMed Links

Gene Details

Gene
MTERF3
Gene Name
mitochondrial transcription termination factor 3
There are no transcripts associated with this gene.

Gene
UQCRB
Gene Name
ubiquinol-cytochrome c reductase binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199975.2 386 Intron NP_001186904.1
NM_001254752.1 386 Silent Mutation CAC,CAT H94H NP_001241681.1
NM_006294.4 386 Intron NP_006285.1

View Full Product Details