Product Details

SNP ID
rs2984531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72352681 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAAACTGCAAGTTTAAATAACA[A/G]ATCTTTATCTTTTCACTATACAACT
Phenotype
MIM: 604761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01504 PubMed Links

Gene Details

Gene
LINC01504
Gene Name
long intergenic non-protein coding RNA 1504
There are no transcripts associated with this gene.

Gene
ZFAND5
Gene Name
zinc finger AN1-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102420.2 4191 UTR 3 NP_001095890.1
NM_001102421.2 4191 UTR 3 NP_001095891.1
NM_001278243.1 4191 UTR 3 NP_001265172.1
NM_001278244.1 4191 UTR 3 NP_001265173.1
NM_001278245.1 4191 UTR 3 NP_001265174.1
NM_006007.3 4191 UTR 3 NP_005998.1

View Full Product Details