Product Details
- SNP ID
-
rs2303555
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:169827714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAGCGGCCGCTGGCCGCGGCTGC[C/T]GGCGGCGAGGACGCGGCGGCGGCCG
- Phenotype
-
MIM: 613831
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
METTL5
PubMed Links
Gene Details
- Gene
- METTL5
- Gene Name
- methyltransferase like 5
There are no transcripts associated with this gene.
- Gene
- UBR3
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 3 (putative)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172070.3 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
NP_742067.3 |
XM_005246299.4 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_005246356.1 |
XM_005246300.4 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_005246357.1 |
XM_006712268.3 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_006712331.1 |
XM_006712269.3 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_006712332.1 |
XM_006712270.3 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_006712333.2 |
XM_006712271.3 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_006712334.1 |
XM_011510604.2 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_011508906.1 |
XM_017003356.1 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_016858845.1 |
XM_017003357.1 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_016858846.1 |
XM_017003358.1 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_016858847.1 |
XM_017003359.1 |
207 |
Silent Mutation |
GCC,GCT |
A69A |
XP_016858848.1 |
View Full Product Details