Product Details

SNP ID
rs2303983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122911499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCGGTTTCTTTTACCTGCACAG[C/T]CGGTGGCCTCCTCCATGCTGGAGGC
Phenotype
MIM: 609298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA5B PubMed Links

Gene Details

Gene
SEMA5B
Gene Name
semaphorin 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031702.3 3520 Missense Mutation GAC,GGC D1028G NP_001026872.2
NM_001256346.1 3520 Missense Mutation GAC,GGC D1028G NP_001243275.1
NM_001256347.1 3520 Missense Mutation GAC,GGC D1082G NP_001243276.1
NM_001256348.1 3520 Missense Mutation GAC,GGC D934G NP_001243277.1
XM_017006638.1 3520 Missense Mutation GAC,GGC D994G XP_016862127.1
XM_017006639.1 3520 Intron XP_016862128.1

View Full Product Details