Product Details
- SNP ID
-
rs3010874
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
62
- Location
-
Chr.1:12716131 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTGAACCATGTCCTAAATGGTTT[A/C]CACTGCGCACAGCTTCCTCTCAGCC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AADACL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80277373] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AADACL3
- Gene Name
- arylacetamide deacetylase like 3
View Full Product Details