Product Details

SNP ID
rs2304145
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:38440938 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAGAATCTTGGGTCCAAAGAAGA[C/G]GGTTCTGGGAGTCTGAAAGGAGGTG
Phenotype
MIM: 180901
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RYR1 PubMed Links

Gene Details

Gene
RYR1
Gene Name
ryanodine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000540.2 Intron NP_000531.2
NM_001042723.1 Intron NP_001036188.1
XM_006723317.2 Intron XP_006723380.1
XM_006723319.2 Intron XP_006723382.1
XM_011527205.2 Intron XP_011525507.1

View Full Product Details