Product Details

SNP ID
rs2304244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48395017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGACAGAGGAAGTTGGCAAGAAG[A/C]CTCGTGGAGGGAGGGGGTTTGAACG
Phenotype
MIM: 602717 MIM: 131235
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GRIN2D PubMed Links

Gene Details

Gene
GRIN2D
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000836.2 Intron NP_000827.2
XM_011526872.1 Intron XP_011525174.1
Gene
KDELR1
Gene Name
KDEL endoplasmic reticulum protein retention receptor 1
There are no transcripts associated with this gene.

View Full Product Details