Product Details

SNP ID
rs2304297
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:42753056 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGCAAGAAAGTCCTCTTTTTCCAT[C/G]TAGCCATCAGTTTTAGCAGATGGGG
Phenotype
MIM: 606888
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHRNA6 PubMed Links

Gene Details

Gene
CHRNA6
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199279.1 1964 UTR 3 NP_001186208.1
NM_004198.3 1964 UTR 3 NP_004189.1

View Full Product Details