Product Details

SNP ID
rs3000857
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:12717928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACCCCATGTTCACAACTAACTT[C/T]CAATTGAAATTCTTCTCATGAGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AADACL3 PubMed Links

Gene Details

Gene
AADACL3
Gene Name
arylacetamide deacetylase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103170.2 Intron NP_001096640.2

View Full Product Details