Product Details

SNP ID
rs2304660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:42767255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGAAGATGGGGAGCTGCTGCCC[A/G]CCCAGGGGCATGGCATCTGGGCTGA
Phenotype
MIM: 604521 MIM: 609050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAAO PubMed Links

Gene Details

Gene
HAAO
Gene Name
3-hydroxyanthranilate 3,4-dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012205.2 2776 UTR 3 NP_036337.2
XM_005264230.3 2776 UTR 3 XP_005264287.1
XM_011532729.2 2776 UTR 3 XP_011531031.1
XM_011532730.2 2776 UTR 3 XP_011531032.1
XM_011532731.2 2776 UTR 3 XP_011531033.1
XM_017003717.1 2776 UTR 3 XP_016859206.1
Gene
MTA3
Gene Name
metastasis associated 1 family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282755.1 2776 Intron NP_001269684.1
NM_001282756.1 2776 Intron NP_001269685.1
NM_020744.3 2776 Intron NP_065795.1
XM_005264456.2 2776 Intron XP_005264513.1
XM_005264458.2 2776 Intron XP_005264515.1
XM_005264459.2 2776 Intron XP_005264516.1
XM_017004561.1 2776 Intron XP_016860050.1
XM_017004562.1 2776 Intron XP_016860051.1
Gene
OXER1
Gene Name
oxoeicosanoid receptor 1
There are no transcripts associated with this gene.

View Full Product Details