Product Details

SNP ID
rs2305122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58545385 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGGTGGGGGCCATAACCTGGGT[C/G]GGAACTTGTAACAGTCTCCCACATC
Phenotype
MIM: 610893 MIM: 601742
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHMP2A PubMed Links

Gene Details

Gene
CHMP2A
Gene Name
charged multivesicular body protein 2A
There are no transcripts associated with this gene.

Gene
MIR6807
Gene Name
microRNA 6807
There are no transcripts associated with this gene.

Gene
TRIM28
Gene Name
tripartite motif containing 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005762.2 Intron NP_005753.1
Gene
ZBTB45
Gene Name
zinc finger and BTB domain containing 45
There are no transcripts associated with this gene.

View Full Product Details