Product Details

SNP ID
rs2305346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76040387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTAGTTTGTTACTAATCCTGCTT[C/T]GTATTCCTCAAAAACCATGGCCCAG
Phenotype
MIM: 604858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRP68 PubMed Links

Gene Details

Gene
SRP68
Gene Name
signal recognition particle 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260502.1 Intron NP_001247431.1
NM_001260503.1 Intron NP_001247432.1
NM_014230.3 Intron NP_055045.2

View Full Product Details