Product Details

SNP ID
rs3021526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97854739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGCCCGGCCGGCCAACCCCTC[C/T]GCCTATGCGGCTGCCTACGCGGGCC
Phenotype
MIM: 602617
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXE1 PubMed Links

Gene Details

Gene
FOXE1
Gene Name
forkhead box E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004473.3 1485 Silent Mutation TCC,TCT S275S NP_004464.2
Gene
PTCSC2
Gene Name
papillary thyroid carcinoma susceptibility candidate 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details