Product Details

SNP ID
rs3024294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44962277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTGCAGGTCAGCTGTGCCAGA[C/G]CTGCTGACTCAGCCGCGATGCTGCA
Phenotype
MIM: 611586
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1QL1 PubMed Links

Gene Details

Gene
C1QL1
Gene Name
complement C1q like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006688.4 Intron NP_006679.1

View Full Product Details