Product Details

SNP ID
rs3024462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:6145530 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGTCTGGGTCTTCACACCTAAG[C/T]CAAAGCAAGAGCTATTTTTGCGTTA
Phenotype
MIM: 134570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F13A1 PubMed Links

Gene Details

Gene
F13A1
Gene Name
coagulation factor XIII A chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000129.3 2411 UTR 3 NP_000120.2

View Full Product Details