Product Details

SNP ID
rs2325971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2845831 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTCTGTCTCAGAATTTGACTAC[C/T]CTAGACTCCTTATGTAAGTGGAGTC
Phenotype
MIM: 608550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCED1A PubMed Links

Gene Details

Gene
PCED1A
Gene Name
PC-esterase domain containing 1A
There are no transcripts associated with this gene.

Gene
VPS16
Gene Name
VPS16, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022575.3 Intron NP_072097.2
NM_080413.2 Intron NP_536338.1

View Full Product Details