Product Details
- SNP ID
-
rs3024826
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:85659042 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTACAGAAAGACAAAAAGAAACC[C/G]CTTTTTATATCTTAACAAAGCAATA
- Phenotype
-
MIM: 178640
MIM: 611594
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SFTPB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147067432] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SFTPB
- Gene Name
- surfactant protein B
- Gene
- USP39
- Gene Name
- ubiquitin specific peptidase 39
There are no transcripts associated with this gene.
View Full Product Details