Product Details

SNP ID
rs2249187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:5056963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTACATTTTCGCGGTCCTGCTAG[C/T]ATGCGTGGTGTTCCACTCTGGCGCC
Phenotype
MIM: 400022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH11Y PubMed Links

Gene Details

Gene
PCDH11Y
Gene Name
protocadherin 11 Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278619.1 Intron NP_001265548.1
NM_032971.2 Intron NP_116753.1
NM_032972.2 Intron NP_116754.1
NM_032973.2 Intron NP_116755.1
XM_017030079.1 Intron XP_016885568.1
XM_017030080.1 Intron XP_016885569.1
XM_017030081.1 Intron XP_016885570.1
XM_017030082.1 Intron XP_016885571.1
XM_017030083.1 Intron XP_016885572.1

View Full Product Details