Product Details

SNP ID
rs2355162
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:158000798 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGTGAAATGAGCTCCTTGCAGGAGA[A/G]AGTAGTTTTATTACTAGAAGATGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UPP2 PubMed Links

Gene Details

Gene
UPP2
Gene Name
uridine phosphorylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135098.1 Intron NP_001128570.1
NM_173355.3 Intron NP_775491.1
XM_005246359.3 Intron XP_005246416.1
XM_017003484.1 Intron XP_016858973.1

View Full Product Details