Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001167740.1 | Intron | NP_001161212.1 | |||
NM_022743.2 | Intron | NP_073580.1 | |||
XM_011544253.1 | Intron | XP_011542555.1 | |||
XM_011544254.2 | Intron | XP_011542556.1 | |||
XM_011544257.1 | Intron | XP_011542559.1 | |||
XM_011544258.2 | Intron | XP_011542560.1 | |||
XM_011544259.2 | Intron | XP_011542561.1 | |||
XM_011544260.1 | Intron | XP_011542562.1 | |||
XM_011544261.2 | Intron | XP_011542563.1 | |||
XM_017002094.1 | Intron | XP_016857583.1 | |||
XM_017002095.1 | Intron | XP_016857584.1 |