Product Details

SNP ID
rs2364160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77508672 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATAAGAAGGAAGCAAAGTGTAT[A/T]ACACACTAGAAAATTCAGCTAGGCT
Phenotype
MIM: 612684 MIM: 605713
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ISM2 PubMed Links

Gene Details

Gene
ISM2
Gene Name
isthmin 2
There are no transcripts associated with this gene.

Gene
SPTLC2
Gene Name
serine palmitoyltransferase long chain base subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004863.3 5489 UTR 3 NP_004854.1
XM_011537384.2 5489 Intron XP_011535686.1

View Full Product Details