Product Details

SNP ID
rs2379240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1283616 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCCTCGCCCAGGGCCAGGCCTC[C/G]GAGACGCCCTGCCCTCCCCTCCCTT
Phenotype
MIM: 602649 MIM: 602756
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf24 PubMed Links

Gene Details

Gene
C19orf24
Gene Name
chromosome 19 open reading frame 24
There are no transcripts associated with this gene.

Gene
CIRBP
Gene Name
cold inducible RNA binding protein
There are no transcripts associated with this gene.

Gene
EFNA2
Gene Name
ephrin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001405.3 800 Intron NP_001396.2
XM_017026449.1 800 UTR 5 XP_016881938.1
XM_017026450.1 800 Intron XP_016881939.1

View Full Product Details