Product Details
- SNP ID
-
rs2379640
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.16:21378777 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGAATCACAGTTGACCTTACCTG[G/T]ATCACCAGCCACTTCCCCAGCTGGA
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNX29P1
PubMed Links
Gene Details
- Gene
- SNX29P1
- Gene Name
- sorting nexin 29 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details