Product Details
- SNP ID
-
rs2385924
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:121615940 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATTCTCTCCTCTGTCTCAATTTT[G/T]CCAGCTCCTTTTTTTAAAAATCACC
- Phenotype
-
MIM: 601636
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HAS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138866924] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAS2
- Gene Name
- hyaluronan synthase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005328.2 |
|
Intron |
|
|
NP_005319.1 |
View Full Product Details