Product Details
- SNP ID
-
rs2407580
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:59305198 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTATCATATTGGCTTTGGGCTTGT[A/C]ATATATAGTCTGTATAATGTTGAGG
- Phenotype
-
MIM: 611370
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FGGY
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80121543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FGGY
- Gene Name
- FGGY carbohydrate kinase domain containing
View Full Product Details