Product Details
- SNP ID
-
rs2408474
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:51757461 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATTTAGCTATAGAAAAAAGGAAA[A/G]AAAAAAATTGTCCTGAGAGCCCACT
- Phenotype
-
MIM: 616163
MIM: 610418
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DHRS12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs542638556] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DHRS12
- Gene Name
- dehydrogenase/reductase 12
- Gene
- WDFY2
- Gene Name
- WD repeat and FYVE domain containing 2
View Full Product Details