Product Details

SNP ID
rs2451376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34025040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGAGATGAGCACAGACCAGCA[C/T]CCTGAGGGCGGGAGCCGAGATTCTA
Phenotype
MIM: 604100
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRM4 PubMed Links

Gene Details

Gene
GRM4
Gene Name
glutamate metabotropic receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000841.3 Intron NP_000832.1
NM_001256809.2 Intron NP_001243738.1
NM_001256811.2 Intron NP_001243740.1
NM_001256812.2 Intron NP_001243741.1
NM_001256813.2 Intron NP_001243742.1
NM_001282847.1 Intron NP_001269776.1
XM_017010790.1 Intron XP_016866279.1
XM_017010791.1 Intron XP_016866280.1
XM_017010792.1 Intron XP_016866281.1
XM_017010793.1 Intron XP_016866282.1

View Full Product Details