Product Details

SNP ID
rs2432688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35582055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCGTTGGACACGCGGCGTCTC[C/T]GGTTGGCAGCTTTGGCGAGCAGCGA
Phenotype
MIM: 602273
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT1 PubMed Links

Gene Details

Gene
GALNT1
Gene Name
polypeptide N-acetylgalactosaminyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020474.3 Intron NP_065207.2
XM_005258239.3 Intron XP_005258296.1
XM_017025692.1 Intron XP_016881181.1
Gene
MIR3975
Gene Name
microRNA 3975
There are no transcripts associated with this gene.

View Full Product Details