Product Details

SNP ID
rs2434041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:15941370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTGAATGCCGAGGGGCGTTTGC[C/T]CAAATTTGGGGAGGGGAAGGATTTG
Phenotype
MIM: 164840 MIM: 605374 MIM: 615968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCN PubMed Links

Gene Details

Gene
MYCN
Gene Name
v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293228.1 Intron NP_001280157.1
NM_001293231.1 Intron NP_001280160.1
NM_001293233.1 Intron NP_001280162.1
NM_005378.5 Intron NP_005369.2
XM_017004168.1 Intron XP_016859657.1
Gene
MYCNOS
Gene Name
MYCN opposite strand
There are no transcripts associated with this gene.

Gene
MYCNUT
Gene Name
MYCN upstream transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details