Product Details

SNP ID
rs2436901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:102552785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTGGAGAAAACTTTCCTTTTCT[A/C]ACTTTTTACATATGCCATTTAGTCT
Phenotype
MIM: 182878
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ODF1 PubMed Links

Gene Details

Gene
ODF1
Gene Name
outer dense fiber of sperm tails 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024410.3 Intron NP_077721.2

View Full Product Details