Product Details

SNP ID
rs2416281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:113514126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGCTGGCAGTCAGGATACCCC[C/T]CTCACCCCAGCGCCCCCCAAACGGC
Phenotype
MIM: 616530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YTHDC2 PubMed Links

Gene Details

Gene
YTHDC2
Gene Name
YTH domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022828.3 Intron NP_073739.3
XM_011543581.1 Intron XP_011541883.1
XM_011543582.2 Intron XP_011541884.1
XM_011543586.2 Intron XP_011541888.1
XM_017009730.1 Intron XP_016865219.1
XM_017009731.1 Intron XP_016865220.1
XM_017009732.1 Intron XP_016865221.1
XM_017009733.1 Intron XP_016865222.1
XM_017009734.1 Intron XP_016865223.1
XM_017009735.1 Intron XP_016865224.1

View Full Product Details